Description
- Filter paper soaked with one or several colored indicators of pH
- Economic range of pH 0 to 14
- Available in a variety of formats to meet individual needs
- Reference chart shows color changes in steps of pH2
Specifications
Specifications
| pH Range | 1 to 11 |
| Format | Roll |
| Quantity | 4 Rolls |
| Type | Grade 911 |
| Length (Metric) | 5 m |
| Width (Metric) | 7 mm |