Learn More
Fisherbrand™ Universal GC Capillary Connectors
For precise connections of fused-silica capillary columns in gas chromatography. Fisherbrand™ Universal GC Capillary Connectors are straight or Y-shaped connectors for joining all columns with an inner diameter of 0.20-0.53mm and an outer diameter of 0.30-0.75mm.
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
Universal Capillary Connectors available in:- Straight Capillary Connector for connecting 2 columns
- Y-Shaped Capillary Connector for connecting 3 columns
Specifications
Universal Capillary Connector | |
Straight | |
10/Box | |
Clear |
0.30 to 0.75 mm | |
Polypropylene box | |
0.20 to 0.53 mm | |
Fused-Silica Capillary Columns in GC |