Learn More
SPEX CertiPrep™ Adjustable Clamp for Geno/Grinder
Shop All SPEX Certiprep Products
Click to view available options
Type:
Adjustable Clamp
Unit Size:
Each
Description
- Fits up to 6 deep-well titer plates and up to 16 tubes of 50mL
- Release mechanism for sliding clamp up and down shaft
Specifications
Specifications
| Type | Adjustable Clamp |
| For Use With (Application) | Geno Grinder |