Description
- Lightweight, reliable polyamide safety gloves
- Polyurethane coating on the inside of the hand and fingertips
- Ideal for precision assembly work
Specifications
Specifications
| Applications | Industrial |
| Coating Material | Polyurethane |
| Color | Gray |
| Exterior Finish | Smooth |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Polyamide |
| Product Type | Mechanical Protection Gloves |
| Size | 11 |
| EN388 Standard | 4141X |
| Show More |