Learn More
Hirschmann™ Union Nut
Product Code. p-7159655
Change view
Click to view available options
Unit Size:
Each
Color:
Red
Turquoise
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Color
10644893 Each -
10004732 Each Turquoise
10710654 Each Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10644893 Supplier Hirschmann™ Supplier No. 9902402
Add to basket
Add to basket
Retainer rings for use with the Pipetus™ pipette filler line.
  • Available in a range of colors
  • Aids in liquid handling
  • Accessory item
Specifications
Type Union Nut
For Use With (Equipment) Pipetus