Learn More
Hirschmann™ Union Nut
Product Code. 10710654
Click to view available options
Color:
Red
Turquoise
Unit Size:
Each
Product Code. 10710654
Brand: Hirschmann™ 9903912
Only null left
Add to basket
Add to basket
Retainer rings for use with the Pipetus™ pipette filler line.
  • Available in a range of colors
  • Aids in liquid handling
  • Accessory item
Specifications
Color Red
Type Union Nut
For Use With (Equipment) Pipetus