Learn More
ABEBA™ Uni6 White Safety Slip-On Shoe S2
Product Code. 16328011
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
16328011 47 1 pair
16308001 35 1 pair
16318001 36 1 pair
16328001 37 1 pair
16338001 38 1 pair
16348001 39 1 pair
16358001 40 1 pair
16368001 41 1 pair
16378001 42 1 pair
16388001 43 1 pair
16398001 44 1 pair
16308011 45 1 pair
16318011 46 1 pair
16338011 48 1 pair
14 options
Product Code. 16328011
Brand: ABEBA™ 31740/47
Only null left
Add to basket
Add to basket
ABEBA™ Uni6 White Safety Slip-On Shoe S2 made of water and chemical resistant microfiber with a slip resistant PU/TPU outsole.
  • Comfort, safety and hygiene
  • Comply to EN ISO 20345 : 2012, S2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Water and chemical resistant microfiber
  • Steel toe cap (200 joules)
  • Instep area with elastic band
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • No side stitching
  • Removable insole Active comfort
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S2 SRC ESD
Color White
Liner Material BreathaBle silver ions lining
Lug Type Padded
Material Microfiber
Size 47
Sole Features Slip and abrasion resistant, ESD dissipative
Sole Material Polyurethane, Thermoplastic Polyurethane
Style Slip-on shoes
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20345 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes