Learn More
ABEBA™ Uni6 Black Safety Trainers S3
Description
- Sporty design and comfort
- Comply to EN ISO 20345 : 2012, S3, SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant PU/TPU outsole
- black functional leather with honeycomb pattern
- Water and abrasion resistent
- Steel toe cap (200 joules)
- Textile antipuncture midsole
- Breathable and antibacterial lining with silver ions
- Sanitized treatment
- Removable insole Active comfort
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20345:2012 S3 SRC ESD |
| Closure Type | Laces |
| Color | Black |
| Eyelets Material | Thermoplastic Polyurethane |
| Liner Material | Breathable silver ions lining |
| Lug Type | Padded |
| Material | Leather |
| Size | 40 |
| Sole Features | Slip and abrasion resistant, ESD dissipative |
| Sole Material | Polyurethane, Thermoplastic Polyurethane |
| Show More |