Learn More
ABEBA™ Uni6 Black Safety Trainers S3
Product Code. 16318041
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
16318041 48 1 pair
16388021 35 1 pair
16398021 36 1 pair
16308031 37 1 pair
16318031 38 1 pair
16328031 39 1 pair
16338031 40 1 pair
16348031 41 1 pair
16358031 42 1 pair
16368031 43 1 pair
16378031 44 1 pair
16388031 45 1 pair
16398031 46 1 pair
16308041 47 1 pair
14 options
Product Code. 16318041
Brand: ABEBA™ 31765/48
Only null left
Add to basket
Add to basket
ABEBA™ Uni6 Black Safety Trainers S3 feature a sporty design and a slip resistant PU/TPU outsole.
  • Sporty design and comfort
  • Comply to EN ISO 20345 : 2012, S3, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • black functional leather with honeycomb pattern
  • Water and abrasion resistent
  • Steel toe cap (200 joules)
  • Textile antipuncture midsole
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable insole Active comfort
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S3 SRC ESD
Closure Type Laces
Color Black
Eyelets Material Thermoplastic Polyurethane
Liner Material Breathable silver ions lining
Lug Type Padded
Material Leather
Size 48
Sole Features Slip and abrasion resistant, ESD dissipative
Sole Material Polyurethane, Thermoplastic Polyurethane
Style Trainer
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20345 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes