Learn More
ABEBA™ Uni6 Black Safety Slip-On Shoe S2
Description
- Comfort, safety and hygiene
- Comply to EN ISO 20345 : 2012, S2, SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant PU/TPU outsole
- Water and chemical resistant microfiber
- Steel toe cap (200 joules)
- Instep area with elastic band
- Breathable and antibacterial lining with silver ions
- Sanitized treatment
- No side stitching
- Removable insole Active comfort
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20345:2012 S2 SRC ESD |
| Color | Black |
| Liner Material | Breathable silver ions lining |
| Lug Type | Padded |
| Material | Microfiber |
| Size | 47 |
| Sole Features | Slip and abrasion resistant, ESD dissipative |
| Sole Material | Polyurethane, Thermoplastic Polyurethane |
| Style | Slip-on shoes |
| Toe Material | Steel |
| Show More |