Learn More
Abeba™ UNI6 6761 Shoes
Product Code. 15652128
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15652128 35 1 pair
15662128 36 1 pair
15672128 37 1 pair
15682128 38 1 pair
15692128 39 1 pair
15602138 40 1 pair
15612138 41 1 pair
15622138 42 1 pair
15632138 43 1 pair
15642138 44 1 pair
15652138 45 1 pair
15662138 46 1 pair
15672138 47 1 pair
15682138 48 1 pair
14 options
Product Code. 15652128
Brand: Abeba™ 6761/35
Only null left
Add to basket
Add to basket
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
  • CE EN ISO 20347:2012 O2 FO SRC
  • Functional leather with honeycomb pattern
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • Water-repellent
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O2 FO SRC
Color Black
Material Leather
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 6761