Learn More
Haldenwanger™ Glazed Porcelain Mortar
Click to view available options
Capacity (Metric):
70 mL
75 mL
100 mL
170 mL
250 mL
400 mL
missing translation for 'unitSize'
Each
Description
- Available in a variety of sizes
- Grinding surfaces are unglazed
- Complies with DIN VDE 0335, type C 100
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Finish | Glazed |
| Diameter (Metric) Inner | 80mm |
| Product Type | Mortar |
| Material | Porcelain |
| Height (Metric) | 50mm |