missing translation for 'onlineSavingsMsg'
Learn More

Undecylenic-10 Acid, Sigma-Aldrich™

Product Code. 15868410
Change view
Click to view available options
Quantity:
500 mL
5 L
Unit Size:
5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15868410 5 L 5 L
15858410 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15868410 Supplier Sigma-Aldrich Supplier No. 8008135000
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-38-9

Specifications

Melting Point 24°C
Density 0.91 g/cm3 at 20°C
Boiling Point 275°C/1013 mbar
Molecular Formula CH2=CH(CH2)8COOH
MDL Number MFCD00004442
Quantity 5 L
Synonym Undecylenic-10 acid, 10-Undecenoic acid, Undecylenic-10 acid
Molecular Weight (g/mol) 184.28
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.