missing translation for 'onlineSavingsMsg'
Learn More

Unc119b Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26201-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894715

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mouse Unc119b is related closely to Unc119, a homolog of C. elegans unc119 protein which was first discovered in C. elegans on the basis of a spontaneous mutation affecting locomotion, feeding behavior and chemosensation. Mouse Unc119b shares 90% sequence identity with human UNC119b protein. UNC119b is a myristoyl-binding protein that acts as a cargo adapter: specifically binds the myristoyl moiety of a subset of N-terminally myristoylated proteins and is required for their localization. Myristoyl-binding activity of UNC119b is required to localize NPHP3 to the primary cilium.
Specifications

Specifications

Unc119b
Polyclonal
Unconjugated
UNC119B
AA407659, AI414892, unc 119 homolog B (C. elegans), Unc119b
Rabbit
Antigen Affinity Chromatography
RUO
106840, 611530, 84747
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A6NIH7, Q8C4B4
UNC119B
Unc119b Fusion Protein Ag24599
150 μL
Primary
Canine, Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers