Learn More
UNC 0631, MedChemExpress
Product Code. 16485868
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16485868 10 mg 10mg
16492017 50 mg 50mg
16473386 25 mg 25mg
16408527 5 mg 5mg
4 options
Product Code. 16485868
Brand: Medchem Express HY13808
MedChemExpress UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.
UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.
Specifications
Chemical Name or Material UNC 0631
CAS 1320288-19-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C37H61N7O2
Quantity 10 mg
Solubility Information DMF : 150 mg/mL (235.88 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (157.25 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (26.21 mM; Need ultrasonic)
SMILES COC1=CC2=C(NC3CCN(CC4CCCCC4)CC3)N=C(N5CCN(C(C)C)CCC5)N=C2C=C1OCCCN6CCCCC6
Molecular Weight (g/mol) 635.93
Formula Weight 635.93
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death