Learn More
Hecht Karl™ Unadjusted Specific Gravity Bottle
Product Code. 10105921
Change view
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10105921 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10105921 Supplier Hecht Karl™ Supplier No. 42570024
  • ONLINE EXCLUSIVE PRICE
    619.51 NOK
    404.36 NOK / Pack of 1
    Save 215.15 NOK
    34% Off
 Restricted Item
Estimated Shipment: 02-03-2027
Specifications
Quantity 1Each
Type Specific Gravity Bottle
Capacity (Metric) 25 mL
Product Type Specific Gravity Bottle