missing translation for 'onlineSavingsMsg'
Learn More

UM-164, MedChemExpress

Product Code. p-200097106
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497307

Brand: Medchem Express HY112182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UM-164 (DAS-DFGO-II) is a highly potent inhibitor of c-Src with a Kd of 2.7 nM. UM-164 also potently inhibits p38α and p38β.

UM-164 (DAS-DFGO-II) is a highly potent inhibitor of c-Src with a Kd of 2.7 nM. UM-164 also potently inhibits p38α and p38β.

Specifications

Chemical Name or Material UM-164
Physical Form Solid
CAS 903564-48-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C30H31F3N8O3S
For Use With (Application) Cancer-Kinase/protease
Synonym DAS-DFGO-II
Solubility Information DMSO : 5 mg/mL (7.80 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CN=C(NC2=NC(C)=NC(N3CCN(CCO)CC3)=C2)S1)NC4=CC(NC(C5=CC=CC(C(F)(F)F)=C5)=O)=CC=C4C
Molecular Weight (g/mol) 640.68
Formula Weight 640.68
Percent Purity 98.91%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.