Learn More
UM-164, MedChemExpress
Product Code. 16497307
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16497307
Brand: Medchem Express HY112182
MedChemExpress UM-164 (DAS-DFGO-II) is a highly potent inhibitor of c-Src with a Kd of 2.7 nM. UM-164 also potently inhibits p38α and p38β.
UM-164 (DAS-DFGO-II) is a highly potent inhibitor of c-Src with a Kd of 2.7 nM. UM-164 also potently inhibits p38α and p38β.
Specifications
Chemical Name or Material UM-164
Physical Form Solid
CAS 903564-48-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C30H31F3N8O3S
For Use With (Application) Cancer-Kinase/protease
Synonym DAS-DFGO-II
Solubility Information DMSO : 5 mg/mL (7.80 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CN=C(NC2=NC(C)=NC(N3CCN(CCO)CC3)=C2)S1)NC4=CC(NC(C5=CC=CC(C(F)(F)F)=C5)=O)=CC=C4C
Molecular Weight (g/mol) 640.68
Formula Weight 640.68
Percent Purity 98.91%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.