Learn More
Biochrom Ultrospec 10 Disposable Cell
Product Code. 11801772 Shop All Harvard Bioscience Products
Change view
Click to view available options
Type:
Disposable Cell
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11801772 Disposable Cell Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11801772 Supplier Biochrom Supplier No. 80200453
  • ONLINE EXCLUSIVE PRICE
    1410.00 NOK
    1292.22 NOK / Pack of 100
    Save 117.78 NOK
    8% Off
Estimated Shipment: 13-01-2027
Ultrospec 10 is a disposable cuvette suited for use in OD600 measurements of cell cultures of bacteria and yeast.
  • For use with Ultrospec 10
  • Made of methacrylate
  • Min. operating volume is 2.5 mL
Specifications
Type Disposable Cell
Material Plastic
Optics OD600
pH Range 5 to 9
Voltage 9 V