Learn More
Merck Ultrafree MC Tubes
Brand:  Merck UFC3000TB
Additional Details : Weight : 25.00000kg
Product Code. 10623402
  • 4955.00 NOK / Pack of 1000
Estimated Shipment: 25-06-2024
Specifications
Clear
Removal of particles and precipitates from aqueous and some solvent-based samples, including protein and nucleic acid solutions
12,000g (1–4 min.)
Filtrate Collection Tube
40 mm
Non-sterile
Polypropylene
1000 Pack
1.5 mL
10.6 mm