Learn More
Sartorius Ultrafiltration Membrane Discs
Product Code. 10245801 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
25 mm
47 mm
63 mm
76 mm
Material (Membrane):
Cellulose
PES
Polysulfone
Thickness (Metric):
120 μm
180 μm
MWCO:
1 kDa
10 kDa
100 kDa
30 kDa
300 kDa
5 kDa
50 kDa
Unit Size:
Pack of 10
Product Code. Diameter (Metric) Material (Membrane) Thickness (Metric) MWCO unitSize
10245801 25 mm Cellulose 180 μm 10 kDa Pack of 10
10022281 47 mm Polysulfone 120 μm 5 kDa Pack of 10
11720945 25 mm Polysulfone 120 μm 5 kDa Pack of 10
15992510 76 mm Polysulfone 120 μm 50 kDa Pack of 10
10173751 25 mm Cellulose 180 μm 5 kDa Pack of 10
11750945 76 mm Polysulfone 120 μm 5 kDa Pack of 10
10123941 47 mm Polysulfone 120 μm 30 kDa Pack of 10
11790945 76 mm Polysulfone 120 μm 10 kDa Pack of 10
10504201 25 mm Cellulose 180 μm 30 kDa Pack of 10
11740945 63 mm Polysulfone 120 μm 5 kDa Pack of 10
10133801 63 mm Cellulose 180 μm 5 kDa Pack of 10
11760945 25 mm Polysulfone 120 μm 10 kDa Pack of 10
11730955 47 mm PES 120 μm 100 kDa Pack of 10
10347261 47 mm Polysulfone 120 μm 1 kDa Pack of 10
10397681 63 mm Polysulfone 120 μm 30 kDa Pack of 10
10337261 47 mm Cellulose 180 μm 5 kDa Pack of 10
15982510 25 mm Polysulfone 120 μm 50 kDa Pack of 10
10748202 47 mm Polysulfone 120 μm 50 kDa Pack of 10
10662602 76 mm Cellulose 180 μm 30 kDa Pack of 10
10439511 76 mm Cellulose 180 μm 10 kDa Pack of 10
10574571 47 mm Cellulose 180 μm 30 kDa Pack of 10
15922520 76 mm Polysulfone 120 μm 300 kDa Pack of 10
10052301 63 mm Cellulose 180 μm 30 kDa Pack of 10
15962510 63 mm Polysulfone 120 μm 1 kDa Pack of 10
10716092 47 mm Polysulfone 120 μm 10 kDa Pack of 10
10032421 76 mm Cellulose 180 μm 5 kDa Pack of 10
10662392 47 mm Polysulfone 120 μm 300 kDa Pack of 10
15972510 76 mm Polysulfone 120 μm 1 kDa Pack of 10
11780945 63 mm Polysulfone 120 μm 10 kDa Pack of 10
15902520 25 mm Polysulfone 120 μm 30 kDa Pack of 10
30 options
Product Code. 10245801
Brand: Sartorius 1443925D
Only null left
Add to basket
Add to basket
For sample concentration in stirred cells
Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.
  • Wide range of MWCO
  • Wide range of disc diameters
  • Compatible with all stirred cells
TRUSTED_SUSTAINABILITY
Specifications
Filter Type Membrane Filter
Format Ultrafiltration Disc
Material Hydrosart
Material (Membrane) Cellulose
MWCO 10 kDa
Sterility Non-sterile
For Use With (Application) For sample concentration in stirred cells
Quantity 10/Pk.
Thickness (Metric) 180 μm
Type Membrane Filter
Diameter (Metric) 25 mm