Learn More
Thermo Scientific™ UltraFast™ UFC-WAX GC Columns
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns.
25812.12 NOK - 30214.55 NOK
Specifications
Product Line | Ultrafast |
---|---|
Stationary Phase | UltraFast UFC-WAX |
For Use With (Equipment) | Trace GC UltraFast Instrument |
For Use With (Application) | GC for Chemicals, GC for Petrochemicals, GC for Environmental Analysis, GC for Flavors and Fragrances |
Length (Metric) | 5 m |
Product Code | Brand | Film Thickness | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Film Thickness | Price | Quantity & Availability | |||||
10594265
|
Thermo Scientific™
UFMC00001010501 |
0.1 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10137934
|
Thermo Scientific™
UFMC00001010503 |
0.2 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns. If your laboratory processes large numbers of samples each day, UltraFast GC columns have the processing speed and throughput you need. These short, narrow-bore columns are designed for use with the Thermo Scientific TRACE GC UltraFast gas chromatograph configuration. Together they deliver a significant gain in analysis speed compared to conventional GC procedures.
Applications:
Chemical
Petrochemical
Environmental
Flavors and fragrances
Tailored Specifically for Robustness and SpeedUltraFast GC columns utilize short (2 to 10m), narrow-bore capillaries and temperature programming conditions usually fast than 2°C/s, which results in peak widths in the 50-200ms range. As a result, analysis times average 1 minute or less. Benefits include:
Dramatically shorter analysis times
Increased sample throughput–by a factor of 20
Longer column lifetimes
UltraFast UFC-WAX Columns uses:
Fatty acid methyl esters (FAMEs)
Essential oils
General
Specifications
Ultrafast | |
Trace GC UltraFast Instrument | |
5 m |
UltraFast UFC-WAX | |
GC for Chemicals, GC for Petrochemicals, GC for Environmental Analysis, GC for Flavors and Fragrances | |
0.1 mm |