Learn More
IKA™ ULTRA-TURRAX™ T 65 Basic Disperser
GREENER_CHOICE
Product Code. 13430597 Shop All IKA Products
Click to view available options
Speed:
7200 rpm
Unit Size:
Each
Product Code. 13430597
Brand: IKA™ 0004023500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides a wide speed range from 8,000 to 30,000rpm that enables users to work at high circumferential speeds even with small rotor diameters. IKA™ ULTRA-TURRAX™ T 65 Basic Disperser offers three rotor-stator configurations for a wide range of applications.
  • Extremely powerful dispersing instrument for volumes from 2 to 50L (H2O) for typical pilot plant stations
  • Reproducible operations due to constant speed even with changes in viscosities
  • Electronic overload protection
  • Cables and plugs not included in the delivery
  • Plug-in connectors facilitate exchange of dispersing elements
  • Stainless steel dispersing elements for use under vacuum and pressure
TRUSTED_SUSTAINABILITY
Specifications
Product Type Basic Disperser
Speed 7200 rpm
Voltage 3 x 400 V
Electrical Requirements 3 x 400 V, 50 Hz, 1800/1500 W
Hertz 50/60 Hz
Wattage 1500/1500 W
Volume Range (Metric) 2 to 50 L
Depth (Metric) 400 mm
Height (Metric) 271 mm
Width (Metric) 185 mm
Model T 65 ULTRA-TURRAX™
Packaging Quantity 1 Ea.
Viscosity 5000 mPas