Learn More
IKA™ ULTRA-TURRAX™ T 18 Digital Disperser
GREENER_CHOICE
Product Code. 13470657 Shop All IKA Products
Click to view available options
Product Type:
Digital Disperser
Digital Disperser Package
Plug Type:
EU Plug
Unit Size:
Each
Product Code. 13470657
Brand: IKA™ 0009022800
Disperses volumes from 1 to 1500mL (H2O) with digital speed display. IKA™ ULTRA-TURRAX™ T 18 Digital Disperser offers a wide speed range from 3000 to 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
Competitively priced dispersing instrument for volumes from 1 to 1500mL (H2O) with digital speed display. Offers a wide speed range from 3000 to 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
  • Digital speed display
  • Electronic speed control
  • Electronic overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Plastic disposable dispersing elements are available in two sizes
  • Error code display
  • Quiet operation
  • Motor protection against overload
  • Digital display for precise monitoring of set and actual speeds 2+1 year after registering at 2+1 www.ika.com/register
  • Rotating knob for adjusting the speed
TRUSTED_SUSTAINABILITY
Specifications
Product Type Digital Disperser Package
Speed 3000 to 25,000 rpm
Voltage 220/240 V
Electrical Requirements 220/240 V, 50/60 Hz, 500 W
Hertz 50 Hz
Wattage 500 W
Volume Range (Metric) 1 mL to 1.5 L
Depth (Metric) 106 mm
Height (Metric) 450 mm
Width (Metric) 87 mm
Display LED
Includes S 18 N - 19 G Dispersing tool
Model T 18 ULTRA-TURRAX™
Packaging Quantity 1 Ea.
Plug Type EU Plug
Temperature Range (Metric) 5°C to 40°
Viscosity 5000 mPas