Learn More
OHAUS™ Ultra Multi-Purpose Clamps
Product Code. 15782299 Shop All Ohaus Products
Change view
Click to view available options
Size (Metric) Grip:
0 to 103 mm
0 to 23 mm
0 to 32 mm
0 to 50 mm
0 to 70 mm
0 to 72 mm
Unit Size:
Each
Product Code. Size (Metric) Grip unitSize
15782299 0 to 23 mm Each
15772299 0 to 32 mm Each
15762299 0 to 50 mm Each
15752299 0 to 70 mm Each
15742299 0 to 72 mm Each
15732299 0 to 103 mm Each
6 options
Product Code. 15782299
Brand: OHAUS™ 30392254
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
OHAUS™ Ultra Multi-Purpose Clamps are designed to securely grip and position a variety of laboratory glassware.
  • Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
  • Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
  • Available as single or dual-adjust in nickel-plated zinc or stainless steel
  • Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
TRUSTED_SUSTAINABILITY
Specifications
Type Multi-purpose Clamp
Material Nickel-plated
Includes Non-slip vinyl sleeves and fiberglass covers
Description Clamp, Multi Purpose, CLM-ULTRA3SZS
Diameter (Metric) 8 mm
Length (English) 6.38 in.
Length (English) Arm 4.000 in.
Length (Metric) 162 mm
Length (Metric) Arm 102 mm
Size Small
Size (Metric) Grip 0 to 23 mm
Jaw Type 3-Prong Single Adjust