Learn More
Syngene™ Ultra Low Particulate Air (ULPA) Filter
Ultra Low Particulate Air (ULPA) Filters are available in various configurations for the containment of particles.
Brand:  Syngene™ SYN-ACFULPA18
Additional Details : Weight : 3.62870kg
 View more versions of this product
Product Code. 17736004
  • 8305.71 NOK / Each
Estimated Shipment: 01-11-2023
Description
  • The filter has a 99.995% efficiency rating for collecting 0.125 micron particles.
Specifications
Specifications
99.9995%
ULPA
No Frame