Learn More
3M™ Nomad™ Ultra Clean Matting 4300
Description
- Disposable, multi-layered contamination control mat
- Designed to help protect clean areas where cleanliness and hygiene are critical factors
- Peel off one layer to expose the next
- Lay onto any clean surface, no edging or holding frame required
- Anti-microbial additive stops the growth of micro-organisms
- Numbered sheets provide control and assists quality assurance audits
Specifications
Specifications
| Dimensions (L x W) | 115 x 90 cm |
| Disposable | Single-use |
| For Use With (Application) | Contamination control |
| Quantity | 40 Sheets/Mat, 6 Mats/Cs. |
| Surface Coating | Pressure Sensitive Adhesive (PSA) |
| Color | Blue |
| Length (English) | 46 in. |
| Length (Metric) | 115 cm |
| Type | Entrance Mat |
| Width (English) | 36 in. |
| Show More |