Learn More
Wesbart™ ULT Freezer Storage Bin
Description
- Storage bins for ULT upright freezers
- For storage of loose items
- All bins are supplied with a handle or hand grip and label holder.
Specifications
Specifications
| Type | Storage Bin |
| Color | Silver |
| For Use With (Application) | Keeping Loose Items Safe Within a Freezer |
| Includes | Handle on One End and a Label Holder |
| Dimensions (L x W x H) Exterior | 610 x 180 x 240 mm |