missing translation for 'onlineSavingsMsg'
Learn More

UKI-1, MedChemExpress

Product Code. 16423008
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16423008 10 mg 10mg
16425747 5 mg 5mg
16424316 100 mg 100mg
16420649 1 mg 1mg
16419576 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16423008 Supplier Medchem Express Supplier No. HY100415
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress UKI-1 (UKI-1C) is a potent urokinase-type plasminogen activator (uPA) inhibitor with a Ki of 0.41 μM. UKI-1 is also a low molecular weight serine protease inhibitor. UKI-1 is a potent antimetastatic agent and inhibits the invasive capacity of carcinoma cells.

UKI-1 (UKI-1C) is a potent urokinase-type plasminogen activator (uPA) inhibitor with a Ki of 0.41 μM. UKI-1 is also a low molecular weight serine protease inhibitor. UKI-1 is a potent antimetastatic agent and inhibits the invasive capacity of carcinoma cells.

Specifications

Chemical Name or Material UKI-1
CAS 220355-63-5
Color White
Purity Grade Notes Research
Molecular Formula C32H47N5O5S
Quantity 10 mg
Synonym UKI-1C
Solubility Information DMSO : 100 mg/mL (162.92 mM; Need ultrasonic)
SMILES O=C(N1CCN(C([C@@H](NS(=O)(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)=O)CC3=CC=CC(C(N)=N)=C3)=O)CC1)OCC
Molecular Weight (g/mol) 613.81
Formula Weight 613.81
Percent Purity 96.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.