missing translation for 'onlineSavingsMsg'
Learn More

UGT2B7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16661-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886354

  • 3468.15 NOK / 150µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The UGTs (EC 2.4.1.17) serve a major role in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B7 has unique specificity for 3,4-catechol estrogens and estriol, suggesting that it may play an important role in regulating the level and activity of these potent estrogen metabolites. Its subcellular location is the microsome.
Specifications

Specifications

UGT2B7
Polyclonal
Unconjugated
UGT2B7
UDPGT 2B7, UDPGT 2B9, UDPGTh 2, UGT2B7, UGT2B9, UGTB2B9
Rabbit
Antigen Affinity Chromatography
RUO
7364
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P16662
UGT2B7
UGT2B7 Fusion Protein Ag10121
150 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers