missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UGT1A6 Polyclonal antibody specifically detects UGT1A6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | UGT1A6 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | GNT1UDPGT 1-6, HLUGP, HLUGP1, MGC29860, Phenol-metabolizing UDP-glucuronosyltransferase, UDP glucuronosyltransferase 1 family, polypeptide A6, UDP glycosyltransferase 1 family, polypeptide A6, UDP-glucuronosyltransferase 1 family polypeptide A6s, UDP-glucuronosyltransferase 1-6, UDP-glucuronosyltransferase 1A6, UDP-glucuronosyltransferase 1-F, UDPGT, UGT1, UGT1%ast;6, UGT1.6, UGT1-06, UGT1A6S, UGT-1F, UGT1FEC 2.4.1.17 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 65-270 of human UGT1A6 (NP_001063.2).,, Sequence:, LLLKESKYYTRKIYPVPYDQEELKNRYQSFGNNHFAERSFLTAPQTEYRNNMIVIGLYFINCQSLLQDRDTLNFFKESKFDALFTDPALPCGVILAEYLGLPSVYLFRGFPCSLEHTFSRSPDPVSYIPRCYTKFSDHMTFSQRVANFLVNLLEPYLFYCLFSKYEELASAVLKRDVDIITLYQKVSVWLLRYDFVLEYPRPVMPN |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?