missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody is specific to UBN1.
Cellular senescence is a hallmark of tumor suppression and tissue aging. Senescent cells contain domains of heterochromatin, called senescence-associated heterochromatin foci (SAHF), that repress proliferation-promoting genes. The protein encoded by this gene binds to proliferation-promoting genes and is required for SAHF formation, enhancing methylation of histone H3.
Specifications
Specifications
| Antigen | UBN1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.29 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | UBN1 |
| Gene Accession No. | Q4G0F8, Q9NPG3 |
| Gene Alias | HIRA binding protein, Protein VT4, ubinuclein 1, UBN1, VT, VT4 |
| Gene Symbols | Ubn1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?