missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Antibody (Ubi-1) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB300-130JF669
This item is not returnable.
View return policy
Description
Ubiquitin Monoclonal antibody specifically detects Ubiquitin in Bovine,C. elegans,Chicken,Drosophila,Equine,Human,Monkey,Mouse,Plant,Porcine,Rat,Zebrafish samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunoblot,Immunocytochemistry/ Immunofluorescence
Specifications
| Ubiquitin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| RPS27A, UBA52, UBB ubiquitin B, UBC | |
| Purified bovine erythrocyte ubiquitin cross-linked with glutaraldehyde to KLH | |
| 0.1 mL | |
| Alzheimers Research, Autophagy, Cancer, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Turnover, Ubiquitin Proteasome Pathway | |
| 6233 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunocytochemistry/Immunofluorescence | |
| Ubi-1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Bovine, C. elegans, Chicken, Drosophila, Equine, Human, Monkey, Mouse, Plant, Pig, Rat, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction