missing translation for 'onlineSavingsMsg'
Learn More

UBE4A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21548-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16891135

  • 1281.92 NOK / 20µL
Estimated Shipment: 14-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ubiquitination requires a ubiquitin-activating enzyme, a ubiquitin-conjugating enzyme, and usually a substrate-specific ubiquitin-protein ligase. Multiubiquitination needs a fourth factor, to bind to the ubiquitin moieties of preformed conjugates and catalyze ubiquitin chain assembly in conjunction with the first three proteins. Human protein KIAA0126, with a predicted 1,073 amino acid residues, is a member of the novel protein family defined by ubiquitination conjugation factor E4. In yeast, E4 activity is linked to cell survival under stress conditions, indicating that eucaryotes use E4-dependent proteolysis pathways for multiple cellular functions.
Specifications

Specifications

UBE4A
Polyclonal
Unconjugated
UBE4A
E4, KIAA0126, UBE4A, UBOX2, UFD2
Rabbit
Antigen Affinity Chromatography
RUO
140630, 315608, 9354
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E9Q735, Q14139, Q6P7A2
Ube4a
UBE4A Fusion Protein Ag16211
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers