missing translation for 'onlineSavingsMsg'
Learn More

UBE2O Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15812-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16895164

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

E2/E3 hybrid ubiquitin-protein ligase that displays both E2 and E3 ligase activities and mediates monoubiquitination of target proteins. Negatively regulates TRAF6-mediated NF-kappa-B activation independently of its E2 activity. Acts as a positive regulator of BMP7 signaling by mediating monoubiquitination of SMAD6, thereby regulating adipogenesis. Mediates monoubiquitination at different sites of the nuclear localization signal (NLS) of BAP1, leading to cytoplasmic retention of BAP1. Also able to monoubiquitinate the NLS of other chromatin-associated proteins, such as INO80 and CXXC1, affecting their subcellular location. Acts as a regulator of retrograde transport by assisting the TRIM27:MAGEL2 E3 ubiquitin ligase complex to mediate ′Lys-63′-linked ubiquitination of WASHC1, leading to promote endosomal F-actin assembly.
Specifications

Specifications

UBE2O
Polyclonal
Unconjugated
UBE2O
E2-230K, KIAA1734, UBE2O, Ubiquitin carrier protein O, Ubiquitin protein ligase O
Rabbit
Antigen Affinity Chromatography
RUO
217342, 63893
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZPJ3, Q9C0C9
UBE2O
UBE2O Fusion Protein Ag8580
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers