missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBE2L3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16578151
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16578151 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16578151 Supplier Invitrogen™ Supplier No. PA592154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RKCGMKNFRN IQVDEANLLT WQGLIVPDNP PYDKGAFRIE INFPAEYPFK PPKITFKTKI YHPNIDEKGQ VCLPVISAEN WKPATKTDQV IQSLIALVND PQPEHPLRAD LAEEYSKDRK KFCKNAEEFT KKYGEKRPVD; Positive Samples: U-251MG, HL-60, K-562, HeLa, Mouse testis, Mouse brain, Mouse skeletal muscle; Cellular Location: Cytoplasm, Nucleus.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L3 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to participate in the ubiquitination of p53, c-Fos, and the NF-kB precursor p105 in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2L3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE2L3
Gene Accession No. P68036, P68037
Gene Alias C79827; E2 ubiquitin-conjugating enzyme L3; E2-F1; L-UBC; UBCE7; UBCH7; UbcM4; Ube2l3; Ubiquitin carrier protein L3; ubiquitin conjugating enzyme E2 L3; ubiquitin conjugating enzyme E2L 3; ubiquitin-conjugating enzyme 7; ubiquitin-conjugating enzyme E2 L3; Ubiquitin-conjugating enzyme E2-F1; ubiquitin-conjugating enzyme E2L 3; ubiquitin-conjugating enzyme UBCH7; ubiquitin-protein ligase L3; Unknown (protein for MGC:127956)
Gene Symbols UBE2L3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 73-212 of human UBE2L3 (NP_0012432841).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22195, 363836, 7332
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.