missing translation for 'onlineSavingsMsg'
Learn More

UBE2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10733-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16897303

  • 1611.84 NOK / 20µL
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2B is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution.
Specifications

Specifications

UBE2B
Polyclonal
Unconjugated
UBE2B
E2 17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6B, UBC2, UBE2B, Ubiquitin carrier protein B, Ubiquitin protein ligase B
Rabbit
Antigen Affinity Chromatography
RUO
22210, 7320, 81816
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P63146, P63147, P63149
UBE2B
UBE2B Fusion Protein Ag1147
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers