Learn More
UBE1 Mouse anti-Human, Unconjugated, Clone: 1H7E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67198-1-IG-20UL
Description
The UBE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBE1. This antibody recognizes human, mouse, rat antigen. The UBE1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| UBE1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1H7E8 | |
| Unconjugated | |
| 20 μL | |
| 7317 | |
| IgG2a |