Learn More
UBE1 Mouse anti-Human, Unconjugated, Clone: 1H7E8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67198-1-IG-20UL
 View more versions of this product
Product Code. 16849705
  • 1710.33 NOK / 20µL
Description
The UBE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBE1. This antibody recognizes human, mouse, rat antigen. The UBE1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
UBE1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
1H7E8
Unconjugated
20 μL
7317
IgG2a