Description
- Calibrated, with constant, for manual measurement
- Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high-temperature or low temperature measurements
- All viscometers are provided with ring marks
- This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
- The recommended minimum flowthrough time is 200 seconds
Specifications
Specifications
| Product Type | Ubbelohde Viscometer |
| Viscosity Range | 20 to 100 cSt |
| Calibrated | Yes |
| Description | 1.03 mm diamter |
| Approximate Constant | 0.1 |
| Certifications/Compliance | ISO 3105, ASTM D 2515, ASTM D 446 |
| Material | Glass |
| Length (Metric) | 285 mm |
| For Use With (Application) | Manual measurements |
| Volume (Metric) | 15 to 20 mL |