Learn More
SI Analytics™ Ubbelohde Viscometers with Additional Tubes
Product Code. 10598811 Shop All SI Analytics Products
Change view
Click to view available options
Approximate Constant:
0.01
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Approximate Constant Unit Size
10598811 0.01 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10598811 Supplier SI Analytics™ Supplier No. 285401933
  • ONLINE EXCLUSIVE PRICE
    8245.00 NOK
    7145.49 NOK / Pack of 1
    Save 1099.51 NOK
    13% Off
In Stock
SI Analytics™ Ubbelohde Viscometers with Additional Tubes feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.
  • Calibrated, with constant, for automatic measurement
  • Ideal for transparent liquids automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high-temperature or low temperature measurements
  • These viscometers are preferably used for automatic measurements when an AVS™ 24 or AVS™ 26 automatic viscometer cleaner is used simultaneously
  • The additional filling and cleaning tube and the glass thread ensure safe operational use
  • The calibrated viscometers are delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
  • The ring marks that are also present serve as auxiliary marks in case the viscometers must be checked by means of manual measurements
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ubbelohde Viscometer
Viscosity Range 1.2 to 10 cSt
Calibrated Yes
Description 0.63 mm diameter
Approximate Constant 0.01
Certifications/Compliance ISO 3105, DIN 51 562, Part 1, BS 133, NFT 60-100
Material Glass
Length (Metric) 290 mm
For Use With (Application) Automatic measurements
Volume (Metric) 18 to 22 mL