Learn More
SI Analytics™ Ubbelohde Viscometer
Product Code. 12028183 Shop All SI Analytics Products
Change view
Click to view available options
Viscosity Range:
0.5 to 3 cSt
0.8 to 5 cSt
1.2 to 10 cSt
10 to 100 cSt
100 to 1000 cSt
1000 to 10,000 cSt
3 to 30 cSt
30 to 300 cSt
5 to 50 cSt
50 to 500 cSt
Approximate Constant:
0.003K
0.005K
0.01K
0.03K
0.05K
0.1K
0.3K
0.5K
10K
1K
Size:
0.47mm Diameter
0.54mm Diameter
0.63mm Diameter
0.84mm Diameter
0.95mm Diameter
1.13mm Diameter
1.50mm Diameter
1.69mm Diameter
2.01mm Diameter
3.60mm Diameter
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Approximate Constant Size Unit Size
12028183 30 to 300 cSt 0.3K 1.50mm Diameter Each
12088173 0.5 to 3 cSt 0.003K 0.47mm Diameter Each
12078173 0.8 to 5 cSt 0.005K 0.54mm Diameter Each
10053940 1.2 to 10 cSt 0.01K 0.63mm Diameter Each
12098173 3 to 30 cSt 0.03K 0.84mm Diameter Each
10701761 5 to 50 cSt 0.05K 0.95mm Diameter Each
12008183 10 to 100 cSt 0.1K 1.13mm Diameter Each
12018183 50 to 500 cSt 0.5K 1.69mm Diameter Each
12068183 1000 to 10,000 cSt 10K 3.60mm Diameter Each
12038183 100 to 1000 cSt 1K 2.01mm Diameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 12028183 Supplier SI Analytics™ Supplier No. 285400259
In Stock
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Ubbelohde Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • The recommended minimum flow through time is 200 seconds
  • Calibrated
  • With constant for automatic measurements
  • Filling quantity of 15 to 20mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Viscosity Range 30 to 300 cSt
Calibrated Yes
Approximate Constant 0.3K
Certifications/Compliance DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
Includes Ring Marks
For Use With (Application) For Determination of Absolute and Relative Kinematic Viscosity of Liquids With Newtonian Flow Behavior
Size 1.50mm Diameter
Volume (Metric) 15 to 20 mL