Learn More
SI Analytics™ Ubbelohde ASTM Viscometer
Product Code. 12028173 Shop All SI Analytics Products
Change view
Click to view available options
Approximate Constant:
0.003K
0.005K
0.01K
0.03K
0.3K
Size:
0.36mm Diameter
0.46mm Diameter
0.58mm Diameter
0.78mm Diameter
1.36mm Diameter
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Approximate Constant Size Unit Size
12028173 0.01K 0.58mm Diameter Each
10400883 0.003K 0.36mm Diameter Each
10099880 0.005K 0.46mm Diameter Each
10588801 0.03K 0.78mm Diameter Each
10508811 0.3K 1.36mm Diameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12028173 Supplier SI Analytics™ Supplier No. 285400534
  • ONLINE EXCLUSIVE PRICE
    5855.00 NOK
    5072.02 NOK / Pack of 1
    Save 782.98 NOK
    13% Off
View all promotions
In Stock
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Ubbelohde ASTM Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • The recommended minimum flow through time is 200 seconds
  • Calibrated with constant for manual measurements
  • Filling quantity of 15 to 20mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Calibrated Yes
Approximate Constant 0.01K
ASTM Numbers D 446
Certifications/Compliance ISO/DIS 3105, ASTM D 2515, ASTM D 446
For Use With (Application) Manual Measurements and Automatic Measurements
Size 0.58mm Diameter