Description
- Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
- The recommended minimum flow through time is 200 seconds
- Calibrated with constant for manual measurements
- Filling quantity of 15 to 20mL
Specifications
Specifications
| Product Type | Viscometer |
| Calibrated | Yes |
| Approximate Constant | 0.03K |
| ASTM Numbers | D 446 |
| Certifications/Compliance | ISO/DIS 3105, ASTM D 2515, ASTM D 446 |
| For Use With (Application) | Manual Measurements and Automatic Measurements |
| Size | 0.78mm Diameter |