Learn More
UBAC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19844-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to UBAC1.
UBAC1 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two RRM domains that bind to RNAs.Specifications
| UBAC1 | |
| Polyclonal | |
| Unconjugated | |
| UBAC1 | |
| GBDR1, KPC2, RP11 432J22.3, UBA domain containing 1, UBAC1, UBADC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10422, 362087, 98766 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XIR9, Q8VDI7, Q9BSL1 | |
| Ubac1 | |
| UBAC1 Fusion Protein Ag13887 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |