Learn More
Ohaus™ U Stand Support Base
Shop All Ohaus Products
Click to view available options
Length (Metric):
178 mm
Unit Size:
Each
Description
- OHAUS offers the largest selection of lab-frame kits, lab-lifts, rods and support stands in the industry to hold glassware safely
- Standard lab-frames or customisable set-ups with options to fit any space or application
- Select from 13 pre-configured lab-frame kits (connectors and feet included) to rods which can be purchased individually and combined with OHAUS accessories to build a customized frame
Specifications
Specifications
| Material | Cast Iron |
| Length (English) | 7 in. |
| Length (Metric) | 178 mm |
| Width (English) | 7 in. |
| Width (Metric) | 178 mm |
| Product Type | Support Stand Base |