Description
- Lightweight, durable disposable NBR gloves
- Texturized fingertips for enhanced grip
- Reliable spray protection when handling chemicals such as acids, alkalis, solids, or aqueous saline solutions
- Silicone-free according to the imprint test
- Touchscreen compatible
- Food safe
Specifications
Specifications
| Color | Blue |
| Disposable | Single-use |
| Exterior Finish | Smooth |
| Latex Free | Yes |
| Material | Nitrile Rubber |
| Palm Thickness (Metric) | 0.08 mm |
| Powdered | Powder Free |
| Product Type | Nitrile Gloves |
| Size | 7 |
| Sterility | Non-sterile |
| Show More |