Learn More
uvex U-Cap Sport Bump Cap - Short Brim
Product Code. 17269221 Shop All Uvex Products
Change view
Click to view available options
Size:
55 to 59 cm
60 to 63 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17269221 55 to 59 cm Each
17279221 60 to 63 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17269221 Supplier uvex Supplier No. 9794403
  • ONLINE EXCLUSIVE PRICE
    302.00 NOK
    290.61 NOK / Pack of 1
    Save 11.39 NOK
    3% Off
View all promotions
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
An innovative bump cap with a hard shell element and honeycomb structure to absorb shock, in addition to greatly reducing the risk of head injuries. Short brim design ensures a wider field of upward vision.
  • Ideal for areas where helmets are not compulsory, but there is still a risk of head injuries
  • Innovative hard shell element in armadillo-design, honeycomb structure, and shock-absorbing elements
  • Reliable impact protection in accordance with EN 812
  • Short brim ensures a wider field of upward vision
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Reusable
Product Type Bump Cap
Size 55 to 59 cm
Includes Armadillo-Style Hard Shell
For Use With (Application) Construction, Machine Operation, Masonry, Painting, and Tunnel Work
Ventilated Vented
Head Style Baseball Cap