Learn More
(+/-)-U-50488 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | U-50488 hydrochloride |
| CAS | 67197-96-0 |
| Molecular Formula | C19 H26 Cl2 N2 O . Cl H |
| Quantity | 10 mg |
| Synonym | Benzeneacetamide','3,4-dichloro-N-methyl-N-[(1R,2R)-2-(1-pyrrolidinyl)cyclohexyl]-','hydrochloride (1:1)','rel-, Benzeneacetamide','3,4-dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]-','monohydrochloride','trans- (9CI), MCV 4133, NIH 9470, U 50488E, U-50488 hydrochloride, rel-3,4-Dichloro-N-methyl-N-[(1R,2R)-2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide monohydrochloride, 3,4-Dichloro-N-methyl-N-[trans-2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride, (±)-U-50488 hydrochloride |
| InChI Formula | InChI=1S/C19H26Cl2N2O.ClH/c1-22(19(24)13-14-8-9-15(20)16(21)12-14)17-6-2-3-7-18(17)23-10-4-5-11-23;/h8-9,12,17-18H,2-7,10-11,13H2,1H3;1H/t17-,18-;/m1./s1 |
| SMILES | Cl.CN([C@@H]1CCCC[C@H]1N2CCCC2)C(=O)Cc3ccc(Cl)c(Cl)c3 |
| IUPAC Name | 2-(3,4-dichlorophenyl)-N-methyl-N-[(1R,2R)-2-pyrrolidin-1-ylcyclohexyl]acetamide;hydrochloride |
| Molecular Weight (g/mol) | 405.79 |
| Formula Weight | 404.1189 |