Learn More
DuPont™ Tyvek™ IsoClean™ Chemo Gown
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 30
Description
- Available in sizes XS to 3X
- Increased skirt length ensures protection against frontal exposure Openness in the back closure increases wearer comfort
- Delivers an ideal balance of protection, durability and comfort
- Made of high density polyethylene using a patented flash spinning process
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based liquid splash
- Manufactured in a controlled environment
Specifications
Specifications
| Size | Small |
| Product Type | Controlled Environments Gown |
| Material | Polyethylene |
| Color | White |
| Closure Type | Hook and Loop |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Cuff Style | Knit |
| Latex Free | Yes |
| Sleeve Style | Long Sleeve |
| Show More |