Description
- Specially designed for use with Tyvek™ apparel
- Flash-spun HDPE construction
- Stitched internal seams
- Front zip closure
- Elasticated face and wrists
- White
- PPE Category I
- Antistatic treatment (EN 1149-5) - on both sides
Specifications
Specifications
| Size | Large |
| Material | HDPE |
| Color | White |
| Closure Type | Zipper |
| Certifications/Compliance | EN 1149-5 |
| Seam Type | Stitched (Internal) |
| Product Type | Hooded Chemical-resistant Jacket |
| For Use With (Application) | Pharmaceutical Handling, Chemical Protection, Lead and Asbestos Abatement/remediation, General Maintenance/operations, Spray Painting and General Clean-up, Amongst Many Others |